(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A833099-100MG A833099-250MG
77256-198EA 52.53 USD
77256-198 77256-200
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene ≥98%, ee 99%
(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₂₈H₃₆FeO₂P₂
MW: 522.38 g/mol
MDL Number: MFCD08561159
CAS Number: 849924-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR