3-Ethoxy-2-fluorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
A833082-10G
A833082-1G
A833082-25G
A833082-5G
77281-598EA
59
USD
77281-598
77281-600
77281-602
77281-604
3-Ethoxy-2-fluorophenylboronic acid 98%
3-Ethoxy-2-fluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀BFO₃ |
MDL Number:
MFCD06798077 CAS Number: 855230-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...