3,5-Dimethoxytoluene 98%

Supplier: Ambeed
Warning

A833055-100G A833055-10G A833055-1G A833055-25G A833055-500G A833055-5G
77281-586EA 167.89 USD
77281-586 77281-588 77281-590 77281-592 77281-594 77281-596
3,5-Dimethoxytoluene 98%
3,5-Dimethoxytoluene

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00015435
CAS Number: 4179-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR