(2-Chloropyridin-4-yl)(cyclopropyl)methanamine 97%

Supplier: Ambeed
Danger

A832240-1G A832240-250MG
77244-402EA 345.05 USD
77244-402 77244-404
(2-Chloropyridin-4-yl)(cyclopropyl)methanamine 97%
(2-Chloropyridin-4-yl)(cyclopropyl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
MDL Number: MFCD18652138
CAS Number: 1270444-14-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR