trans-1-(Boc-amino)-4-cyanocyclohexane 97%

Supplier: Ambeed
Warning

A831402-1G A831402-250MG A831402-100MG
77268-460EA 165 USD
77268-460 77268-462 77749-622
trans-1-(Boc-amino)-4-cyanocyclohexane 97%
trans-1-(Boc-amino)-4-cyanocyclohexane

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₂
MDL Number: MFCD11707221
CAS Number: 873537-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR