6-(Aminomethyl)nicotinonitrile dihydrochloride 98%

Supplier: Ambeed
Warning

A831352-100MG A831352-250MG
77268-456EA 111.24 USD
77268-456 77268-458
6-(Aminomethyl)nicotinonitrile dihydrochloride 98%
6-(Aminomethyl)nicotinonitrile dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂N₃
MW: 206.07 g/mol
MDL Number: MFCD30471639
CAS Number: 1350362-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR