(3-Chlorophenyl)acetic acid methyl ester 98%

Supplier: Ambeed
Warning

A831255-100G A831255-1G A831255-25G A831255-5G
77281-576EA 114 USD
77281-576 77281-578 77281-580 77281-582
(3-Chlorophenyl)acetic acid methyl ester 98%
(3-Chlorophenyl)acetic acid methyl ester

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOCH₃
MDL Number: MFCD00800679
CAS Number: 53088-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR