4-Cyanobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A831044-10G A831044-1G A831044-25G A831044-5G
77293-932EA 30 USD
77293-932 77293-934 77293-936 77293-938
4-Cyanobenzenesulfonyl chloride 98%
4-Cyanobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClNO₂S
MDL Number: MFCD00052337
CAS Number: 49584-26-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR