4-Bromo-2-thiophenecarbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromothiophene-2-carbonitrile, 4-Bromo-2-cyanothiophene

A830905-1G A830905-5G A830905-25G
77293-928EA 11.33 USD
77293-928 77293-930 77749-621
4-Bromo-2-thiophenecarbonitrile 98%
4-Bromo-2-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₂BrNS
MW: 188.0500 g/mol
MDL Number: MFCD00114951
CAS Number: 18791-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR