4-Hydroxy-2,6-dimethylbenzoic acid 95%

Supplier: Ambeed
Warning

A830604-100MG A830604-1G A830604-250MG A830604-5G
77293-918EA 25.75 USD
77293-918 77293-920 77293-922 77293-924
4-Hydroxy-2,6-dimethylbenzoic acid 95%
4-Hydroxy-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00963146
CAS Number: 75056-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR