2,5-Dimethoxyaniline 98%

Supplier: Ambeed
Warning

A830577-100G A830577-25G A830577-500G A830577-5G A830577-1000G
77256-188EA 14 USD
77256-188 77256-190 77256-192 77256-194 77747-588
2,5-Dimethoxyaniline 98%
2,5-Dimethoxyaniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD00008368
CAS Number: 102-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR