2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine Hydrochloride, 3-Chloro-5-(trifluoromethyl)picolinylamine hydrochloride, [3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methylamine hydrochloride

A830438-100MG A830438-250MG A830438-5G
77244-376EA 11 USD
77244-376 77244-378 77244-380
2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride 95%
2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₆ClF₃N₂·ClH
MDL Number: MFCD00210134
CAS Number: 326476-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR