4-Methylthiobenzamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylbenzothioamide, Thio-p-toluamide

A830366-1G A830366-25G A830366-5G
77256-182EA 11.33 USD
77256-182 77256-184 77256-186
4-Methylthiobenzamide 98%
4-Methylthiobenzamide

Precaución: for Research Use Only

Formula: C₈H₉NS
MW: 151.23 g/mol
MDL Number: MFCD00173750
CAS Number: 2362-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR