4-Bromopyridine-2,6-dicarbaldehyde 97%

Supplier: Ambeed
Danger

A829694-100MG A829694-1G
77293-914EA 84 USD
77293-914 77293-916
4-Bromopyridine-2,6-dicarbaldehyde 97%
4-Bromopyridine-2,6-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₂
MW: 214.01 g/mol
MDL Number: MFCD09910218
CAS Number: 128184-01-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR