1-Methyl-4-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

A829062-1G A829062-250MG A829062-5G
77244-368EA 43.26 USD
77244-368 77244-370 77244-372
1-Methyl-4-nitro-1H-indazole 98%
1-Methyl-4-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.1600 g/mol
MDL Number: MFCD09607929
CAS Number: 26120-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR