Methyl-1-naphthoate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Naphthalenecarboxylic acid methyll ester, 1-Naphthoic acid methyll ester, Methyl 1-Naphthalenecarboxylate, Methyl 1-naphthoate

A828999-100G A828999-25G
77244-364EA 399.64 USD
77244-364 77244-366
Methyl-1-naphthoate 95%
Methyl 1-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.2066 g/mol
MDL Number: MFCD00094243
CAS Number: 2459-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR