3-Hydroxy-ɑ-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(1-Hydroxyethyl)phenol, 1-(3-Hydroxyphenyl)ethanol

A828398-10G A828398-1G A828398-25G A828398-5G
77281-566EA 135.96 USD
77281-566 77281-568 77281-570 77281-572
3-Hydroxy-ɑ-methylbenzyl alcohol 98%
3-Hydroxy-alpha-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00238621
CAS Number: 2415-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR