1,3-Bis(trimethylsilyloxy)propane 95% (by GC)

Supplier: Ambeed
Warning

Synonyms: 2,2,8,8-Tetramethyl-3,7-dioxa-2,8-disilanonane

A827985-1G A827985-25G A827985-5G
77256-176EA 19.57 USD
77256-176 77256-178 77256-180
1,3-Bis(trimethylsilyloxy)propane 95% (by GC)
1,3-Bis(trimethylsilyloxy)propane

Precaución: for Research Use Only

Formula: C₉H₂₄O₂Si₂
MW: 220.46 g/mol
MDL Number: MFCD00161340
CAS Number: 17887-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR