Boc-d-homoserine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-HoSer-OH

A827960-1G A827960-250MG
77244-356EA 33.99 USD
77244-356 77244-358
Boc-d-homoserine 97%
Boc-d-homoserine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.235 g/mol
MDL Number: MFCD12545888
CAS Number: 745011-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR