5-Aminobenzene-1,2,3-tricarboxylic acid 96%

Supplier: Ambeed
Warning

A827650-1G A827650-25G A827650-5G
77268-426EA 54 USD
77268-426 77268-428 77268-430
5-Aminobenzene-1,2,3-tricarboxylic acid 96%
5-Aminobenzene-1,2,3-tricarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₆
MDL Number: MFCD03839869
CAS Number: 37141-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR