5-Chloro-1,3,3-trimethyl-2-methyleneindoline 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2,3-dihydro-1,3,3-trimethyl-2-methylene-1H-indole

A827547-100G A827547-10G A827547-1G A827547-25G
77281-558EA 338.87 USD
77281-558 77281-560 77281-562 77281-564
5-Chloro-1,3,3-trimethyl-2-methyleneindoline 95%
5-Chloro-1,3,3-trimethyl-2-methyleneindoline

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClN
MW: 207.70 g/mol
MDL Number: MFCD00005814
CAS Number: 6872-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR