3,5-Dibromo-1-isopropyl-1H-1,2,4-triazole 97%

Supplier: Ambeed
Warning

A827421-1G A827421-250MG
77244-352EA 309 USD
77244-352 77244-354
3,5-Dibromo-1-isopropyl-1H-1,2,4-triazole 97%
3,5-Dibromo-1-isopropyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₅H₇Br₂N₃
MW: 268.93 g/mol
MDL Number: MFCD16811118
CAS Number: 1240567-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR