Methyl-4-(methylamino)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(methylamino)benzoate

A827114-10G A827114-1G A827114-25G A827114-5G
77256-148EA 48.41 USD
77256-148 77256-150 77256-152 77256-154
Methyl-4-(methylamino)benzoate 98%
Methyl-4-(methylamino)benzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00017198
CAS Number: 18358-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR