3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-β-Glu-OH
A826943-100MG
A826943-250MG
A826943-1G
77281-554EA
48
USD
77281-554
77281-556
77595-314
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 97%
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid
Precaución: for Research Use Only
Formula:
C₂₀H₁₉NO₆ |
MDL Number:
MFCD03002700 CAS Number: 247217-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...