2-((2-(4-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)phenoxy)ethyl)(methyl)amino)acetic acid hydrochloride 98%

Supplier: Ambeed
Warning

A826662-10MG A826662-1MG A826662-25MG A826662-2MG A826662-50MG A826662-5MG
77268-414EA 142.14 USD
77268-414 77268-416 77268-418 77268-420 77268-422 77268-424
2-((2-(4-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)phenoxy)ethyl)(methyl)amino)acetic acid hydrochloride 98%
2-((2-(4-(Benzo[d][1,3]dioxol-5-yl)-2-(tert-butyl)phenoxy)ethyl)(methyl)amino)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₈ClNO₅
MW: 421.91 g/mol
MDL Number: MFCD12828768
CAS Number: 1779796-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR