2-(4-Ethyl-3-iodophenyl)-2-methylpropanoic acid 98%

Supplier: Ambeed
Warning

A826661-100G A826661-5G
77244-348EA 163 USD
77244-348 77244-350
2-(4-Ethyl-3-iodophenyl)-2-methylpropanoic acid 98%
2-(4-Ethyl-3-iodophenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅IO₂
MDL Number: MFCD22571222
CAS Number: 1256584-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR