5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A825911-100MG A825911-5G
77268-412EA 58.71 USD
77268-412 77595-312
5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde 97%
5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.559 g/mol
MDL Number: MFCD16619816
CAS Number: 117007-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR