2-Amino-5-methylphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-methylphenyboronic acid, pinacol ester, 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A825909-1G A825909-250MG A825909-5G
77256-134EA 155.53 USD
77256-134 77256-136 77256-138
2-Amino-5-methylphenylboronic acid pinacol ester 98%
2-Amino-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
MDL Number: MFCD06795664
CAS Number: 948592-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR