3-Chloro-2-methylbenzonitrile 98%

Supplier: Ambeed
Danger

A825409-10G A825409-1G A825409-25G A825409-5G
77281-536EA 75.19 USD
77281-536 77281-538 77281-540 77281-542
3-Chloro-2-methylbenzonitrile 98%
3-Chloro-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.60 g/mol
MDL Number: MFCD00045598
CAS Number: 54454-12-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR