(S)-(-)-[(S)-2-Diphenylphosphinoferrocenyl](N,N-dimethylamino)(2-diphenylphosphinophenyl)methane ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R)-1-[(S)-α-(Dimethylamino)-2-(diphenylphosphino)benzyl]-2-diphenylphosphinoferrocene, TANIAPHOS
A825292-100MG
A825292-1G
A825292-250MG
A825292-50MG
77256-128EA
51
USD
77256-128
77256-130
77256-132
77747-576
(S)-(-)-[(S)-2-Diphenylphosphinoferrocenyl](N,N-dimethylamino)(2-diphenylphosphinophenyl)methane ≥99%
(S)-(-)-[(S)-2-Diphenylphosphinoferrocenyl](N,N-dimethylamino)(2-diphenylphosphinophenyl)methane
Precaución: for Research Use Only
Formula:
C₄₃H₃₉FeNP₂ |
MDL Number:
MFCD04117709 CAS Number: 850444-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...