1,3-Difluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A825023-100G A825023-25G A825023-5G
77293-898EA 283.25 USD
77293-898 77293-900 77293-902
1,3-Difluoro-5-nitrobenzene 98%
1,3-Difluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
MDL Number: MFCD00012142
CAS Number: 2265-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR