2-Chloroethyl benzoate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzoic Acid 2-Chloroethyl Ester
A824649-100G
A824649-25G
A824649-5G
77268-404EA
99.91
USD
77268-404
77268-406
77268-408
2-Chloroethyl benzoate 99%
2-Chloroethyl benzoate
Precaución: for Research Use Only
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol |
MDL Number:
MFCD00045290 CAS Number: 939-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...