(11aR)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine 98%

Supplier: Ambeed
Warning

A823900-1G A823900-250MG
77268-400EA 607.7 USD
77268-400 77268-402
(11aR)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine 98%
(11aR)-1,11-Bis(diphenylphosphino)dibenzo[d,f][1,3]dioxepine

Precaución: for Research Use Only

Formula: C₃₇H₂₈O₂P₂
MW: 566.56 g/mol
CAS Number: 301847-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR