3-Aminopyrazine-2-carbohydrazide 97%

Supplier: Ambeed
Warning

A823746-10G A823746-1G A823746-250MG
77281-516EA 157.59 USD
77281-516 77281-518 77281-520
3-Aminopyrazine-2-carbohydrazide 97%
3-Aminopyrazine-2-carbohydrazide

Precaución: for Research Use Only

Formula: C₅H₇N₅O
MW: 153.14 g/mol
MDL Number: MFCD00115105
CAS Number: 6761-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR