6-(Methoxycarbonyl)nicotinic acid 98%

Supplier: Ambeed
Warning

A823308-1G A823308-5G
77268-388EA 27 USD
77268-388 77573-626
6-(Methoxycarbonyl)nicotinic acid 98%
6-(Methoxycarbonyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD09702182
CAS Number: 17874-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR