4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: DAO-IN-2

A823229-1G A823229-250MG A823229-25G
77293-894EA 100.94 USD
77293-894 77293-896 77749-609
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₂S
MW: 167.19 g/mol
MDL Number: MFCD07368542
CAS Number: 39793-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR