1-Ethyl-3-methylimidazolium iodide 99+%

Supplier: Ambeed
Danger

A823006-25G A823006-5G
77244-336EA 21 USD
77244-336 77244-338
1-Ethyl-3-methylimidazolium iodide 99+%
1-Ethyl-3-methylimidazolium iodide

Precaución: for Research Use Only

Formula: C₆H₁₁IN₂
MDL Number: MFCD03701101
CAS Number: 35935-34-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR