3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R)-(6-methoxyquinolin-4-yl)(5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione ≥98%, ee 99%

Supplier: Ambeed
Warning

A822984-100MG A822984-1G A822984-250MG A822984-5G
77256-120EA 92.7 USD
77256-120 77256-122 77256-124 77256-126
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R)-(6-methoxyquinolin-4-yl)(5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione ≥98%, ee 99%
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R)-(6-methoxyquinolin-4-yl)(5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione

Precaución: for Research Use Only

Formula: C₃₂H₂₈F₆N₄O₃
MW: 630.58 g/mol
MDL Number: MFCD30802181
CAS Number: 1256245-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR