6-Fluoro-2-methylpyrido[3,4-d]pyrimidin-4(3H)-one 97%

Supplier: Ambeed
Danger

A822782-100MG A822782-1G
77268-384EA 184.37 USD
77268-384 77268-386
6-Fluoro-2-methylpyrido[3,4-d]pyrimidin-4(3H)-one 97%
6-Fluoro-2-methylpyrido[3,4-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₆FN₃O
MW: 179.15 g/mol
MDL Number: MFCD30189089
CAS Number: 2089325-37-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR