2-Phenylethyl cinnamate 99%

Supplier: Ambeed
Warning

Synonyms: Phenethyl cinnamate, Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester

A822534-100G A822534-25G
77281-506EA 17.51 USD
77281-506 77281-508
2-Phenylethyl cinnamate 99%
2-Phenylethyl cinnamate

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₂
MW: 252.31 g/mol
MDL Number: MFCD00022050
CAS Number: 103-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR