4-Methyl-2-(p-tolyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methyl-2-(4-methylphenyl)pyridine
A822477-100MG
A822477-1G
A822477-250MG
77293-888EA
59.74
USD
77293-888
77293-890
77293-892
4-Methyl-2-(p-tolyl)pyridine 97%
4-Methyl-2-(p-tolyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₃H₁₃N MW: 183.25 g/mol |
MDL Number:
MFCD09031945 CAS Number: 80635-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...