GNE-3511 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-[[6-(3,3-difluoropyrrolidin-1-yl)-4-[1-(oxetan-3-yl)piperidin-4-yl]pyridin-2-yl]amino]pyridine-4-carbonitrile, 2-((6-(3,3-Difluoropyrrolidin-1-yl)-4-(1-(oxetan-3-yl)piperidin-4-yl)pyridin-2-yl)amino)isonicotinonitrile

A822343-10MG A822343-1MG A822343-25MG A822343-5MG
77268-376EA 132.87 USD
77268-376 77268-378 77268-380 77268-382
GNE-3511 ≥98%
GNE-3511

Precaución: for Research Use Only

Formula: C₂₃H₂₆F₂N₆O
MW: 440.49 g/mol
MDL Number: MFCD28502215
CAS Number: 1496581-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR