(3aS,8aS)-(+)-4,4,8,8-Tetrakis(3,5-di-i-propylphenyl)tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo[4,5-e]dioxaphosphepin 98%

Supplier: Ambeed
Warning

A822283-100MG A822283-1G A822283-250MG
77256-114EA 85.49 USD
77256-114 77256-116 77256-118
(3aS,8aS)-(+)-4,4,8,8-Tetrakis(3,5-di-i-propylphenyl)tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo[4,5-e]dioxaphosphepin 98%
(3aS,8aS)-(+)-4,4,8,8-Tetrakis(3,5-di-i-propylphenyl)tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo[4,5-e]dioxaphosphepin

Precaución: for Research Use Only

Formula: C₆₁H₈₁O₄P
MW: 909.26 g/mol
MDL Number: MFCD23704800
CAS Number: 1422371-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR