3-Nitro-o-toluidine 98%

Supplier: Ambeed
Danger

A821928-100G A821928-25G A821928-5G
77281-494EA 65 USD
77281-494 77281-496 77281-498
3-Nitro-o-toluidine 98%
3-Nitro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00007731
CAS Number: 603-83-8
UN: 2660
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR