GSK-4112 98+%

Supplier: Ambeed
Warning

Synonyms: 1,1-Dimethylethyl-N-[(4-chlorophenyl)methyl]-N-[(5-nitro-2-thienyl)methyl])glycinate, GSK4112, Tert-butyl 2-((4-chlorobenzyl)((5-nitrothiophen-2-yl)methyl)amino)acetate, SR 6452

A820682-10MG A820682-1MG A820682-50MG A820682-5MG
77244-320EA 29 USD
77244-320 77244-322 77244-324 77244-326
GSK-4112 98+%
GSK-4112

Precaución: for Research Use Only

Formula: C₁₈H₂₁ClN₂O₄S
MW: 396.89 g/mol
MDL Number: MFCD12912412
CAS Number: 1216744-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR