(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-3-Cl-Ala-OMe·HCl

A820181-100G A820181-25G A820181-500G
77281-488EA 63.86 USD
77281-488 77281-490 77281-492
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 98%
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO₂·ClH
MW: 174.026 g/mol
MDL Number: MFCD00066119
CAS Number: 112346-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR