Mesylate[(di(1-adamantyl)-n-butylphosphine)-2-(2'-amino-1,1'-biphenyl)]palladium(II) 98%

Supplier: Ambeed
Warning

A819748-100MG A819748-5G A819748-1G A819748-250MG
77268-370EA 35 USD
77268-370 77268-372 77552-516 77618-298
Mesylate[(di(1-adamantyl)-n-butylphosphine)-2-(2'-amino-1,1'-biphenyl)]palladium(II) 98%
Mesylate[(di(1-adamantyl)-n-butylphosphine)-2-(2'-amino-1,1'-biphenyl)]palladium(II)

Precaución: for Research Use Only

Formula: C₃₈H₅₄NO₃PPdS
MDL Number: MFCD22200553
CAS Number: 2230788-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR