2,4-Diamino-6-butylamino-1,3,5-triazine 98+%

Supplier: Ambeed
Warning

Synonyms: N2-Butyl-1,3,5-triazine-2,4,6-triamine, N2-Butylmelamine, 2-Butylamino-4,6-diamino-1,3,5-triazine

A819598-1G A819598-5G A819598-25G
77281-480EA 10.3 USD
77281-480 77281-482 77749-607
2,4-Diamino-6-butylamino-1,3,5-triazine 98+%
2,4-Diamino-6-butylamino-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₇H₁₄N₆
MW: 182.23 g/mol
MDL Number: MFCD00059764
CAS Number: 5606-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR