7-Iodoquinazolin-2-amine 97%
Supplier: Ambeed
|
Warning
|
A819438-100MG
A819438-250MG
77268-366EA
52.53
USD
77268-366
77268-368
7-Iodoquinazolin-2-amine 97%
7-Iodoquinazolin-2-amine
Precaución: for Research Use Only
Formula:
C₈H₆IN₃ MW: 271.05 g/mol |
MDL Number:
MFCD29761922 CAS Number: 1935610-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...