(S)-N-Cbz-4-Amino-2-hydroxybutyric acid 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-HABA-OH

A819432-100G A819432-10G A819432-1G A819432-5G A819432-25G
77281-472EA 480 USD
77281-472 77281-474 77281-476 77281-478 77569-526
(S)-N-Cbz-4-Amino-2-hydroxybutyric acid 97%
(S)-N-Cbz-4-Amino-2-hydroxybutyric acid

Precaución: For research use only.

Formula: C₁₂H₁₅NO₅
MDL Number: MFCD00209957
CAS Number: 40371-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR